Learn More
Medchemexpress LLC HIF-1α-IN-2 hydrochloride | 98.1% | 397.92 | 10 MG
SDP
Catalog No. 5000438606
Encompass_Preferred
HIF-1α-IN-2 hydrochloride is an effective HIF-1α inhibitor with anticancer potencies (IC50s of 28 nM and 15 nM in MDA-MB-231 and MiaPaCa-2 cells, respectively). It suppresses HIF-1α expression by blocking transcription and protein translation.
  • Effective HIF-1α inhibitor
  • Suppresses HIF-1α expression by blocking transcription and protein translation
  • Anticancer potencies (IC50s of 28 nM and 15 nM in MDA-MB-231 and MiaPaCa-2 cells)
Catalog No. 50-004-38606 Supplier Medchemexpress LLC Supplier No. HY115903A10MG
HIF-1α-IN-2 hydrochloride is an effective HIF-1α inhibitor with anticancer potencies (IC50s of 28 nM and 15 nM in MDA-MB-231 and MiaPaCa-2 cells, respectively). It suppresses HIF-1α expression by blocking transcription and protein translation.
  • Effective HIF-1α inhibitor
  • Suppresses HIF-1α expression by blocking transcription and protein translation
  • Anticancer potencies (IC50s of 28 nM and 15 nM in MDA-MB-231 and MiaPaCa-2 cells)