Learn More
Cayman Chemical HIF-1 (C-Term) Polyclonal Antibody, 1 ea
SDP
Supplier:  Cayman Chemical 100064211500
Encompass_Preferred
Antigen: peptide from the C-terminal region of human HIF-16 Host: rabbit Species Reactivity: (+) human, mouse, and simian HIF-1 Applications: IHC, WB HIF-1 is a transcription factor that accumulates under low-oxygen conditions. Following hypoxic stimulus and cytoplasmic accumulation, HIF-1 migrates to the nucleus where, with other transcription factors, it drives the production of stress-adaptive proteins. This response is essential for maintenance of normal oxidative physiology, however overexpression in cancer cells promotes tumor survival.
Catalog No. NC0400565