Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HIF-2α-IN-2 | 1672666-82-8 | 99.9% | 50 MG
SDP

Catalog No. 5000338725
Encompass_Preferred

HIF-2α-IN-2 is an inhibitor of hypoxia-inducible factors (HIF-2α), with an IC50 of 16 nM in a scintillation proximity assay (SPA). It was originally extracted from patent WO2015035223A1 (Compound 232).

  • Inhibits hypoxia-inducible factors (HIF-2α)
  • Features an IC50 of 16 nM in SPA
  • Molecular formula: C17H13F2NO4S
  • Molecular weight: 365.35
  • Appears as a white to off-white solid

Catalog No. 50-003-38725 Supplier Medchemexpress LLC Supplier No. HY13026450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HIF-2α-IN-2 is an inhibitor of hypoxia-inducible factors (HIF-2α), with an IC50 of 16 nM in a scintillation proximity assay (SPA). It was originally extracted from patent WO2015035223A1 (Compound 232).

  • Inhibits hypoxia-inducible factors (HIF-2α)
  • Features an IC50 of 16 nM in SPA
  • Molecular formula: C17H13F2NO4S
  • Molecular weight: 365.35
  • Appears as a white to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.