Learn More
Medchemexpress LLC HY-130264 5mg Medchemexpress, HIF-2α-IN-2 CAS:1672666-82-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1302645MG
Encompass_Preferred
Medchemexpress, HY-130264 5mg HIF-2α-IN-2 CAS:1672666-82-8 HIF-2α-IN-2 is a hypoxia-inducible factors (HIF-2α) inhibitor extracted from patent WO2015035223A1, Compound 232, has an IC50 of 16 nM in scintillation proximity assay (SPA). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1234