Learn More
Medchemexpress LLC HIF-2α-IN-4 | 882268-69-1 | 100.0% | 287.32 | 2.87 MG
SDP
Catalog No. 5000348149
Encompass_Preferred
HIF-2α-IN-4 is a potent inhibitor of hypoxia inducible factor-2α (HIF-2α) translation. It has an IC50 of 5 μM and decreases both constitutive and hypoxia-induced HIF-2α protein expression. This compound links its 5'UTR iron-responsive element to oxygen sensing.
  • Potent inhibitor of hypoxia inducible factor-2α (HIF-2α) translation
  • Decreases both constitutive and hypoxia-induced HIF-2α protein expression
  • Links its 5'UTR iron-responsive element to oxygen sensing
  • IC50 of 5 μM
Catalog No. 50-003-48149 Supplier Medchemexpress LLC Supplier No. HY13674810MM/1ML
Add to Cart
Add to Cart
HIF-2α-IN-4 is a potent inhibitor of hypoxia inducible factor-2α (HIF-2α) translation. It has an IC50 of 5 μM and decreases both constitutive and hypoxia-induced HIF-2α protein expression. This compound links its 5'UTR iron-responsive element to oxygen sensing.
  • Potent inhibitor of hypoxia inducible factor-2α (HIF-2α) translation
  • Decreases both constitutive and hypoxia-induced HIF-2α protein expression
  • Links its 5'UTR iron-responsive element to oxygen sensing
  • IC50 of 5 μM