Learn More
TARGETMOL CHEMICALS INC HIF1-IN-3 5MG
SDP
Catalog No. 502578940
Encompass_Preferred
Also available in 2 mg 10 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HIF1-IN-3 (Benzenepropanamide N-[(8-hydroxy-7-quinolinyl)(4-methoxyphenyl)methyl]-) is an effective inhibitor of HIF-1 (EC50 = 0.9 uM) and can be used in anticancer studies. purity: 99%
Catalog No. 50-257-8940 Supplier TARGETMOL CHEMICALS INC Supplier No. T98905MG
Also available in 2 mg 10 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HIF1-IN-3 (Benzenepropanamide N-[(8-hydroxy-7-quinolinyl)(4-methoxyphenyl)methyl]-) is an effective inhibitor of HIF-1 (EC50 = 0.9 uM) and can be used in anticancer studies. purity: 99%