Learn More
Meso Scale Discovery HIF1alpha WCL20ate
SDP
Catalog No. 5000078256
Encompass
The HIF-1alpha Whole Cell Lysate Kit from Meso Scale Discovery is used to detect HIF-1alpha protein in cell lysates The kit contains a 96-well plate coated with capture antibody a detection antibody and buffers and reagents needed to perform the assay The kit can detect endogenous levels of HIF-1alpha from mammalian cell lysates with high sensitivity and without the need for transfection Results are obtained in just a few hours using electrochemiluminescence detection on the MSD platform The kit provides a fast sensitive and high-throughput method to monitor HIF-1alpha protein expression
Catalog No. 50-000-78256 Supplier Meso Scale Discovery Supplier No. K150DKD3
Add to Cart
Add to Cart
Request a Quote
The HIF-1alpha Whole Cell Lysate Kit from Meso Scale Discovery is used to detect HIF-1alpha protein in cell lysates The kit contains a 96-well plate coated with capture antibody a detection antibody and buffers and reagents needed to perform the assay The kit can detect endogenous levels of HIF-1alpha from mammalian cell lysates with high sensitivity and without the need for transfection Results are obtained in just a few hours using electrochemiluminescence detection on the MSD platform The kit provides a fast sensitive and high-throughput method to monitor HIF-1alpha protein expression