Learn More
Meso Scale Discovery HIF1alpha Whole Cell Lysate
SDP
Catalog No. 5000081700
Encompass
The HIF-1alpha Whole Cell Lysate Kit from Mesoscale Discovery provides reagents to detect HIF-1alpha protein levels in cell lysates The kit contains a 96-well plate coated with capture antibody detection antibody read buffer sample diluent and lyophilized recombinant HIF-1alpha standard to generate a standard curve This immunoassay allows sensitive and accurate measurement of HIF-1alpha levels with as little as 15 g of total protein per well The electrochemiluminescent detection method provides a wide dynamic range and high sensitivity with low background This product enables quantification of HIF-1alpha protein expression in cell lysates for hypoxia and cancer research
Catalog No. 50-000-81700 Supplier Meso Scale Discovery Supplier No. K150DKD2
The HIF-1alpha Whole Cell Lysate Kit from Mesoscale Discovery provides reagents to detect HIF-1alpha protein levels in cell lysates The kit contains a 96-well plate coated with capture antibody detection antibody read buffer sample diluent and lyophilized recombinant HIF-1alpha standard to generate a standard curve This immunoassay allows sensitive and accurate measurement of HIF-1alpha levels with as little as 15 g of total protein per well The electrochemiluminescent detection method provides a wide dynamic range and high sensitivity with low background This product enables quantification of HIF-1alpha protein expression in cell lysates for hypoxia and cancer research