Learn More
Aviva Systems Biology Corp HIF1AN Antibody - N-terminal region (ARP39316_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39316_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HIF1AN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HIF1AN. Alternate names: FIH1ApplicationWB. Species reactivity: Horse, Human, Pig, Rabbit
Catalog No. 50-014-56981