Learn More
Aviva Systems Biology Corp HIF1AN Antibody - middle region (ARP39317_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39317_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HIF1AN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HIF1AN. Alternate names: DKFZp762F1811, FIH1, FLJ20615, FLJ22027ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56982