Learn More
Roche Diagnostics KAPA HiFi HotStart Uracil+ ReadyMix Kit (250 rnx), 250 x 50 uL reactions
Catalog No. 501965264 Shop All Roche Diagnostics Products
Encompass
KAPA HiFi HotStart Uracil+ ReadyMix (2X) contains DNA Polymerase, reaction buffer, dNTPs and MgCl2 (at a final concentration of 2.5 mM). *This item is non returnable.
This item is not returnable. View return policy
Catalog No. 50-196-5264 Supplier Roche Diagnostics Supplier No. 07959079001
KAPA HiFi HotStart Uracil+ ReadyMix (2X) contains DNA Polymerase, reaction buffer, dNTPs and MgCl2 (at a final concentration of 2.5 mM). *This item is non returnable.