Learn More
Azzota Corp High-Frequency Adjustable Power Ultrasonic Cleaners, Benchtop, 53 Khz, 22.5 L, 500W, 20C-60C, Indies Size: 50X15X30Cm, 1-199 Min.
Catalog No. 501612781
Encompass_Preferred
High-frequency Adjustable Power Ultrasonic Cleaners, Benchtop, 53 kHz, 22.5 L, 500W, 20C-60C, Indies size: 50x15x30cm, 1-199 Min.
Catalog No. 50-161-2781 Supplier Azzota Corp Supplier No. BHA500T
High-frequency Adjustable Power Ultrasonic Cleaners, Benchtop, 53 kHz, 22.5 L, 500W, 20C-60C, Indies size: 50x15x30cm, 1-199 Min.