Learn More
Azzota Corp Ultrasonic Cleaner, Model Bhf500, 53 Khz Frequency, 22.5L Capacity, 500 W, 1-99 Min. Timer, Lcd Display, With Drain
Catalog No. 501612786
Encompass_Preferred
Ultrasonic Cleaner, Model BHF500, 53 kHz Frequency, 22.5L Capacity, 500 W, 1-99 Min. Timer, LCD Display, with Drain
Catalog No. 50-161-2786 Supplier Azzota Corp Supplier No. BHF500
Ultrasonic Cleaner, Model BHF500, 53 kHz Frequency, 22.5L Capacity, 500 W, 1-99 Min. Timer, LCD Display, with Drain