Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics™ by AquaPhoenix Hig Copper Comparator
SDP

Catalog No. 13299671
Change view
Click to view available options
Range:
1 to 10 ppm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
13-299-671 1 to 10 ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-299-671 Supplier CHEMetrics™ by AquaPhoenix Supplier No. C3510
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Copper comparator designed for comparison against color formed during the reaction. Supports visual analysis of copper test results.

  • C-3510 High Range Copper Comparator is a component of K-3510 Copper (soluble) CHEMets Visual Test Kit and is designed to be compared against the color formed in R-3510 Copper CHEMets Refill.
  • The comparator's range is applicable for many drinking water, water treatment, and aquaculture applications. Color standards are yellow in color and cover a range of 0-10 ppm copper (Cu).

Specifications

pH Range 7
Volume (Metric) 0.5 mL
For Use With (Equipment) K to 3510
Range 1 to 10 ppm
Type Copper Comparator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.