Learn More
Leica Biosystems Hig-profile disp.blades DB80HS
Supplier:  Leica Biosystems 14035843491
Encompass_Preferred
When your application calls for thicker sections, look to the high-profile disposable blades Leica 818 with sharpness, longevity, and consistency.The rectangular shaped corners allow the microtomist to use the full length of the microtome blade for sectioning. The dispenser alignment pin allows direct loading into suitable blade holders.For use with Leica Biosystems microtomes classified and labeled as medical devices for histological medical diagnosis, e.g. cancer diagnosis.For in Vitro Diagnostic Use
Catalog No. 50-255-2453