Learn More
ATCC HIG-82; Synoviocyte; Rabbit (oryctolagus cuniculus)
Catalog No. 502383478
Encompass
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
HIG-82 is a synoviocyte cell line that was isolated from the synovium of a knee joint of a 6-month-old, female rabbit. This cell line was deposited by HI Georgescu, CH Evans.
Catalog No. 50-238-3478 Supplier ATCC Supplier No. CRL1832
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
HIG-82 is a synoviocyte cell line that was isolated from the synovium of a knee joint of a 6-month-old, female rabbit. This cell line was deposited by HI Georgescu, CH Evans.