Learn More
Medchemexpress LLC Higenamine hydrochloride | 11041-94-4 | 307.77 | 1 ML
SDP
Catalog No. 5000416082
Encompass_Preferred
Higenamine hydrochloride (HY-N2037A) is a selective LSD1 inhibitor, isolated from aconite, demonstrating anti-inflammatory and antibacterial activities. It attenuates IL-1β-induced apoptosis through the ROS-mediated PI3K/Akt signaling pathway, protects brain cells from oxygen deprivation, and promotes bone formation in osteoporosis via the SMAD2/3 pathway. This compound is widely used in research for cancer, inflammation, cardiorenal syndrome, and other diseases.
  • Selective LSD1 inhibitor
  • Exhibits anti-inflammatory activity
  • Exhibits antibacterial activity
  • Attenuates IL-1β-induced apoptosis
  • Protects brain cells from oxygen deprivation
  • Promotes bone formation in osteoporosis
Catalog No. 50-004-16082 Supplier Medchemexpress LLC Supplier No. HYN2037A10MM/1ML
Higenamine hydrochloride (HY-N2037A) is a selective LSD1 inhibitor, isolated from aconite, demonstrating anti-inflammatory and antibacterial activities. It attenuates IL-1β-induced apoptosis through the ROS-mediated PI3K/Akt signaling pathway, protects brain cells from oxygen deprivation, and promotes bone formation in osteoporosis via the SMAD2/3 pathway. This compound is widely used in research for cancer, inflammation, cardiorenal syndrome, and other diseases.
  • Selective LSD1 inhibitor
  • Exhibits anti-inflammatory activity
  • Exhibits antibacterial activity
  • Attenuates IL-1β-induced apoptosis
  • Protects brain cells from oxygen deprivation
  • Promotes bone formation in osteoporosis