Learn More
Medchemexpress LLC Higenamine hydrochloride | 11041-94-4 | 307.77 | 500 MG
SDP
Catalog No. 5000414586
Encompass_Preferred
Higenamine hydrochloride is a selective inhibitor of LSD1 with an IC50 of 1.47 μM, naturally sourced from aconite. It exhibits anti-inflammatory and antibacterial properties and can mitigate IL-1β-induced apoptosis via the ROS-mediated PI3K/Akt signaling pathway. Additionally, it offers protective effects for brain cells against oxygen deprivation and stimulates bone formation in osteoporosis through the SMAD2/3 pathway. It is utilized in research for various conditions, including cancer, inflammation, and cardiorenal syndrome.
  • Selective LSD1 inhibitor (IC50=1.47 μM)
  • Derived from aconite
  • Possesses anti-inflammatory activity
  • Exhibits antibacterial activity
  • Attenuates IL-1β-induced apoptosis
  • Protects brain cells from oxygen deprivation
  • Promotes bone formation in osteoporosis
  • Relevant for studying cancer, inflammation, and cardiorenal syndrome
Catalog No. 50-004-14586 Supplier Medchemexpress LLC Supplier No. HYN2037A500MG
Higenamine hydrochloride is a selective inhibitor of LSD1 with an IC50 of 1.47 μM, naturally sourced from aconite. It exhibits anti-inflammatory and antibacterial properties and can mitigate IL-1β-induced apoptosis via the ROS-mediated PI3K/Akt signaling pathway. Additionally, it offers protective effects for brain cells against oxygen deprivation and stimulates bone formation in osteoporosis through the SMAD2/3 pathway. It is utilized in research for various conditions, including cancer, inflammation, and cardiorenal syndrome.
  • Selective LSD1 inhibitor (IC50=1.47 μM)
  • Derived from aconite
  • Possesses anti-inflammatory activity
  • Exhibits antibacterial activity
  • Attenuates IL-1β-induced apoptosis
  • Protects brain cells from oxygen deprivation
  • Promotes bone formation in osteoporosis
  • Relevant for studying cancer, inflammation, and cardiorenal syndrome