Learn More
McMaster-Carr Panel-Mount Flowmeter with Flow-Control Knob
Catalog No. NC2219674
Encompass
Panel-Mount Flowmeter with Flow-Control Knob 303 Stainless Steel Fittings, 1/4 NPT Female, 0.25 gpm to 2.5 gpm
This item is not returnable. View return policy
Catalog No. NC2219674 Supplier McMaster-Carr Supplier No. 2968K31
Panel-Mount Flowmeter with Flow-Control Knob 303 Stainless Steel Fittings, 1/4 NPT Female, 0.25 gpm to 2.5 gpm