Learn More
Lascar Electronics Inc High Accuracy Temp & Relative Humidty Data Logger with LCD Display
SDP
Catalog No. NC1856450
Encompass_Preferred
This standalone data logger measures over 16,000 readings over a -35 to +80°C (-31 to +176°­­F) and 0 to 100% humidity (RH) range. At the touch of a button, use the on-board display to cycle between the current, minimum and maximum temperature logs seen during the session for both temperature and humidity. Easily set up the logger and view downloaded data by plugging the unit into a PC’s USB port and using the free EasyLog software provided. Data can then be graphed, printed and exported to other applications for detailed analysis. Supplied with ½ AA battery and wall mount clip.
Catalog No. NC1856450 Supplier Lascar Electronics Inc Supplier No. ELUSB2LCD+
This standalone data logger measures over 16,000 readings over a -35 to +80°C (-31 to +176°­­F) and 0 to 100% humidity (RH) range. At the touch of a button, use the on-board display to cycle between the current, minimum and maximum temperature logs seen during the session for both temperature and humidity. Easily set up the logger and view downloaded data by plugging the unit into a PC’s USB port and using the free EasyLog software provided. Data can then be graphed, printed and exported to other applications for detailed analysis. Supplied with ½ AA battery and wall mount clip.