Learn More
Bachem H-Tyr-cyclo(-D-Dab-Arg-Phe-Phe-D-Trp-Lys-Thr-Phe)
SDP
Supplier:  Bachem 4041674.0001
Encompass_Preferred
Tyr-(D-Dab4,Arg5,D-Trp8)-cyclo-Somatostatin-14 (4-11) Trifluoroacetate, H-Tyr-cyclo(-D-Dab-Arg-Phe-Phe-D-Trp-Lys-Thr-Phe), 496849-46-8, C67H85N15O11, Mr 1276.51, 1mg. KE 108. High affinity ligand for all five somatostatin receptors (sst1-sst5). In AtT-20 mouse anterior pituitary tumor cells which mainly express sst2 and sst5 receptors the binding affinity of this somatostatin analog was found to be higher than that of somatostatin-14 and octreotide. In a model of epileptogenesis, however, KE 108 did not exhibit antiepileptic activity suggesting that the high potency and efficacy of a synthetic ligand to all known somatostatin receptors may not reproduce entirely the effects of the natural ligand.
Catalog No. 50-194-6727