Learn More
Genscript Corporation High Affinity Ni-Charged Resin
Supplier:  Genscript Corporation L0066625
Encompass
GenScript High Affinity Ni-Charged Resin FF is an 6% highly cross-linked agarose medium covalently coupled to a chelating agent that binds Ni2 by four coordination sites for high-affinity purification of polyhistidine-tagged recombinant proteins
Catalog No. 50-000-57415