Learn More
Genscript Corporation High Affinity Ni-Charged Resin
Catalog No. 5000058649
Encompass
GenScript High Affinity Ni-Charged Resin FF is an 6% highly cross-linked agarose medium covalently coupled to a chelating agent that binds Ni2 by four coordination sites for high-affinity purification of polyhistidine-tagged recombinant proteins
Catalog No. 50-000-58649 Supplier Genscript Corporation Supplier No. L006665
GenScript High Affinity Ni-Charged Resin FF is an 6% highly cross-linked agarose medium covalently coupled to a chelating agent that binds Ni2 by four coordination sites for high-affinity purification of polyhistidine-tagged recombinant proteins