Learn More
Genscript Corporation High Affinity Ni-Charged Resin
Supplier:  Genscript Corporation L0022325
Encompass
GenScript High Affinity Ni-Charged Resin is an 4% cross-linked agarose medium covalently coupled to a chelating agent that binds Ni2+ by four coordination sites for high-affinity purification of polyhistidine-tagged recombinant proteins. High Affinity Ni-Charged Resin has low Ni2+ leakage, high protein-binding capacity and stability, and is compatible with a wide range of additives used in protein purification. This makes High Affinity Ni-Charged Resin the excellent choice for high performance purification of polyhistidine-tagged proteins.
Catalog No. 50-274-421