Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One High Altitude Coagulation Tubes with Sodium Citrate Solution
GSA_VA

Catalog No. p-10000001
Change view
Click to view available options
Capacity (Metric):
1.8 mL
3.15 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
22-040-053 1.8 mL
22-040-055 3.15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-040-053 Supplier Greiner Bio-One Supplier No. 454323
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

For routine hemostasis testing

  • For use in high altitude elevations above 5000 ft.
  • 13 x 75 mm tubes filled with buffered tri-sodium citrate solution
  • 3.2 % (0.109 mol/L) concentration
  • Mixing ratio is one part citrate to nine parts blood when tube is filled appropriately as indicated by fill mark on label
  • Clear as glass with equal or better performance
  • Molded to be virtually unbreakable, providing a better level of safety than glass
  • 18-month shelf life from date of manufacture
  • Blue nonscrew safety pull cap can be removed with a gentle pull

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type High Altitude Sodium Citrate Blood Collection Tube
Additive 3.2% Citrate Solution
Material PET
Capacity (Metric) 1.8 mL
Closure Color Blue with White Ring
Tube Size 13 x 75 mm
For Use With (Application) High altitude elevations above 5000 ft.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.