Learn More
Corning™ 1536-Well, Cell Culture-Treated, Flat-Bottom, High Base Microplate
GREENER_CHOICE
Catalog No. 07202707 Shop All Corning Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
07-202-707 Black
1 options
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Catalog No. 07-202-707 Supplier Corning™ Supplier No. 4705
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Color:Black; Sterility:Sterile; Lid:Without Lid
  • No logo or lettering for improved heat sealing and ease of liquid handling
  • Raised well bottoms for higher sensitivity
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Superior flatness minimizes variability within a plate and between plates
  • Flat bottom wells are compatible with automated dispensers including the PerkinElmer FlexDrop™
  • Barcoded
TRUSTED_SUSTAINABILITY
Specifications
Barcode Includes Barcode
Lid Without Lid
Array 32 x 48
Color Black
Material Cycloolefin Copolymer
No. of Wells 1536
Volume (Metric) Working Up to 8 μL
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
No. per Case 50
No. per Pack 10
Quantity 50/Cs.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov