Learn More
Corning™ High Base Cyclic Olefin Copolymer Microplate
GREENER_CHOICE
Catalog No. 07202702 Shop All Corning Products
Change view
Click to view available options
Color:
White
Catalog No. Color
07-202-702 White
1 options
Catalog No. 07-202-702 Supplier Corning™ Supplier No. 4372
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Raised well bottoms for higher sensitivity. The High Base Cyclic Olefin Copolymer Microplate has excellent signal dynamic range.
Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.
  • 127 μm film thickness
  • 536-well low base, black/clear or white/clear bottom microplates
  • Bar coded
  • Custom bar codes available for compatibility with the Kalypsys system and with UHTS systems
  • Low auto-fluorescence
  • Broad chemical resistance including DMSO and alcohol
  • High mechanical stability
  • Optimized for flatness and uniformity
  • Low birefringence
TRUSTED_SUSTAINABILITY
Specifications
Barcode Includes Barcode
Array 32 x 48
Binding Type Not Treated
Color White
Material Cycloolefin Copolymer
No. of Wells 1536
Volume (Metric) Working Up to 8 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov