Learn More
VEOLIA WTS Oxidizer Cartridge, High-capacity
Catalog No. NC0654142
Encompass
High-capacity, 300-mL, Oxidizer Reagent Cartridge for the Sievers M-Series. Replace after 3 months of use.
This item is not returnable. View return policy
Catalog No. NC0654142 Supplier VEOLIA WTS Supplier No. APF 9030002
High-capacity, 300-mL, Oxidizer Reagent Cartridge for the Sievers M-Series. Replace after 3 months of use.