Learn More
Analytical Sales & Services High Capacity Rupture Proof Diaphragm Damper
Catalog No. 5000805088
Encompass
High Capacity Rupture Proof Diaphragm Damper\n\n• 316 SS and PEEK\n• Standard 10-32 Outlet Ports (Maximum Operating Pressure, 200 PSI)\n\nQty/Pkg: Each
Catalog No. 50-008-05088 Supplier Analytical Sales & Services Supplier No. 592100
High Capacity Rupture Proof Diaphragm Damper\n\n• 316 SS and PEEK\n• Standard 10-32 Outlet Ports (Maximum Operating Pressure, 200 PSI)\n\nQty/Pkg: Each