Learn More
Fisherbrand™ DuraPorter™ XL High-Capacity Sample Transport Tote
Catalog No. 17183973 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
Red
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-183-973 Yellow
17-183-974 Red
17-183-975 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-183-973
Add to Cart
Add to Cart
Reliable solution for the safe and convenient transportation of laboratory samples, instruments, and valuable items. DuraPorter™ XL are crafted to meet the stringent demands of clinical and research laboratories, this transport container ensures the protection and preservation of your cargo.
  • Built to Last and Designed with the User in Mind
  • Sturdy and spacious, ideal for transporting a large volume of samples
  • The transparent lid enables effortless identification of contents, reducing the risk of contamination
  • Equipped with articulated handles featuring silicone grips for comfortable and secure handling
  • Transparent design helps comply with OSHA 29 CFR 1910.1030 standards without having to re-label containers and enabling the user to visibly check sample integrity and meets UN3373 secondary packaging transport requirements
  • Emphasizing Security and Safety
  • A silicone seal ensures a tight enclosure, safeguarding samples from most environmental elements
  • Dual latches with security tag feature guarantee a secure closure, providing tamper-evidence and ensuring the integrity and authenticity of your samples
  • Exceptional Versatility
    • Includes a MultiTube Rack Insert to accommodate various tube sizes. Additionally, two other insert options for blood tubes up to 10 mL and general research tubes up to 15 mL are available separately
    • The bottom compartment is generously sized, allowing for carrying most tube racks, instruments or supplies according to your specific requirements
  • Stainless Steel, PP, Elastomeric Compound, ABS
Specifications
Product Type Specimen Transport Tote
Closure Included Yes
Graduated No
Material Stainless Steel
Shape Rectangular
Sterility Non-sterile
Autoclavable Not Autoclavable
Capacity (Metric) 19 L
Closure Type Latched Lid
Color Yellow
Graduations None
Fit for Purpose Certified Quality