Learn More
Mopec High Capacity Section Dryer
SDP
Catalog No. 22445404 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Microscope Slide Dryer
Catalog No. Product Type
22-445-404 Microscope Slide Dryer
1 options
Catalog No. 22-445-404 Supplier Mopec Supplier No. BK701
Add to Cart
Add to Cart
The High Capacity Section Dryer is an ideal solution for high workload laboratories. Similar to the Mini Section Dryer, it’s designed to efficiently and rapidly blow temperature-controlled air through the base of the instrument to dry specimens without the risk of overheating.
  • Fan-assisted heating dramatically reduces drying time
  • Designed to take up minimal bench-top space
  • Digital temperature control
  • Rapid heating system ensures system is ready to use in less time
  • Anti-microbial coating to help prevent bacteria and virus growth
  • Timer Units (BK701-T1 and BK701-T2) allow monitoring of four slide racks each and ensures that each rack has the correct amount of time in the dryer
Specifications
Product Type Microscope Slide Dryer
For Use With (Equipment) Specimens
Dimensions (L x W x H) 17.5 x 17 x 22.5 in.