Learn More
StatLab High Capacity Slide Dryer 4-Rack Timer Unit 1-4
SDP
Catalog No. 21010363
Change view
Click to view available options
Slide Capacity:
390
Catalog No. Slide Capacity
21-010-363 390
1 options
This item is not returnable. View return policy
Catalog No. 21-010-363 Supplier StatLab Supplier No. SISDRTMR1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Allows monitoring of the first through fourth slide racks when used with the High Capacity Slide Dryer (SI-SDR-HC).
  • Ensures that each slide rack has exactly the correct time in the drying chamber, indicates visually and audibly when an individual slide rack has completed drying
  • Intended for use with the StatLab High Capacity Slide Dryer (SI-SDR-HC), sold separately
  • Additional timer (SI-SDR-TMR2) available for maximum capacity
Specifications
Product Type Slide Dryer Timer
Slide Capacity 390