Learn More
StatLab High Capacity Slide Dryer 8-Slide Rack Holder
SDP
Catalog No. 21060046
Change view
Click to view available options
Product Type:
Slide Holder Rack
Catalog No. Product Type
21-060-046 Slide Holder Rack
1 options
Catalog No. 21-060-046 Supplier StatLab Supplier No. SISDRHLDR
Add to Cart
Add to Cart
Used to identify slide racks when used with the 4 Rack Timer Units (1-4) and (5-8) on the High Capacity Slide Dryer (SI-SDR-HC).
  • Each slide rack entry is numbered which corresponds to the rack number on the 4 Rack LEAN Timer ensuring that each slide rack has exactly the correct time in the drying chamber
  • Used to identify slide racks when used with the 4 Rack Timer Units (1-4) and (5-8) on the StatLab High Capacity Slide Dryer (SI-SDR-HC)
  • Compatible with slide racks from market leading staining instruments; Leica, Sakura and Thermo Scientific
  • Made from stainless steel
  • Contains one rack
Specifications
Product Type Slide Holder Rack