Learn More
Malvern Panalytical High Conc Zeta Potential Cell
Catalog No. NC0475448
Encompass
High concentration zeta potential cell kit. To enable the zeta potential measurement of more concentrated samples than the folded capillary cell or dip cell. Software version 6.20 or later is required. Software is included.
Catalog No. NC0475448 Supplier Malvern Panalytical Supplier No. ZEN1010
Add to Cart
Add to Cart
Request a Quote
High concentration zeta potential cell kit. To enable the zeta potential measurement of more concentrated samples than the folded capillary cell or dip cell. Software version 6.20 or later is required. Software is included.