Learn More
Diagnostic Biosystems High Contrast DAB (Liquid Format)
Supplier:  Diagnostic Biosystems K055
Encompass_Preferred
3,3'-Diaminobenzidine (DAB), is a widely used chromogen for immunoperoxidase staining, and is well accepted among pathologists because of its increased sensitivity and decreased background when compared to amino ethylcarbazole (AEC). Specimens stained with DAB can be dehydrated, cleared, and permanently mounted. High Contrast DAB is more sensitive and stable than traditional working DAB solutions. High Contrast DAB offers several noteworthy improvements and benefits as compared to traditional working DAB solutions. High Contrast DAB is highly sensitive, providing the cost-effective option of further diluting the primary antibody. Being stable for 2 weeks (as opposed to 6 hours for traditional DAB working solutions), High Contrast DAB allows the user the convenience of making one working solution for the entire week’s work. Hazardous waste generation from unused DAB solution is also significantly reduced.
Catalog No. 50-240-7592