Learn More
*AGAROSE BEAD TECHNOLOGIES High Den Glyoxal 4 Rapid Run™-25mL
Catalog No. NC1755563
Encompass_Preferred
High Density Glyoxal 4 Rapid Run™ allows a covalent binding of agarose to lysine amino groups. This product is adequate to work in large scale process (High Pressure). The resin has a high amount of aldehydes groups (40-60 µmoles Glyoxal /ml gel) providing multiple binding points and High immobilized ligand stability. The resin has been manufactured using 4% Rapid Run™ agarose beads, because of this the pore size is adequate to work with enzymes, proteins & antibodies and the stability at high flow is adequate to large scale process.Tech Specs:Description : High Flow. Multiple Binding points. High immobilized ligand stability. Exclusion limit: ~3×10^7Activation degree (μmol glyoxyl/ml gel) : 40-60
Catalog No. NC1755563 Supplier *AGAROSE BEAD TECHNOLOGIES Supplier No. 4RRGH125
High Density Glyoxal 4 Rapid Run™ allows a covalent binding of agarose to lysine amino groups. This product is adequate to work in large scale process (High Pressure). The resin has a high amount of aldehydes groups (40-60 µmoles Glyoxal /ml gel) providing multiple binding points and High immobilized ligand stability. The resin has been manufactured using 4% Rapid Run™ agarose beads, because of this the pore size is adequate to work with enzymes, proteins & antibodies and the stability at high flow is adequate to large scale process.Tech Specs:Description : High Flow. Multiple Binding points. High immobilized ligand stability. Exclusion limit: ~3×10^7Activation degree (μmol glyoxyl/ml gel) : 40-60