Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ High Duster XF with Hand Grip
GREENER_CHOICE

Catalog No. 17000334 Shop All Contec Inc. Products
Change view
Click to view available options
Length (English):
24.25 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
17-000-334 24.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-000-334 Supplier Contec™ Supplier No. MGMP0008
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The High Duster XF makes cleaning hard-to-reach spaces easier when paired with MicroGenesis Microfiber High Duster Covers. The flexible body of the high duster tool bends to hug surface contours and reduces operator strain.

  • Flexible frame reaches areas where typical dusters cannot
  • Reduces cross contamination risk when paired with a Microfiber High Duster Cover

Specifications

Product Type Duster Tool
Flammable No
Length (English) 24.25 in.
Length (Metric) 62 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.