Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dentec Safety™ Comfort-Air™ Series 400 Elastomeric Facepiece With P100 Filter Assembly
SDP

Catalog No. 19222467 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
Size:
Medium/Large
Small/Medium
Packaging Type:
Clamshell
Display Bag
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Packaging Type
19-222-467 Small/Medium Display Bag
19-222-468 Medium/Large Display Bag
19-222-473 Small/Medium Clamshell
19-222-474 Medium/Large Clamshell
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-222-467 Supplier Dentec Safety™ Supplier No. 4SMLP0ZAM
Add to Cart
Add to Cart

Half-mask elastomeric respirator with P100 filter assembly is ideal when used in welding, dust or oil mist applications.

  • Crafted from elastomeric rubber material and complete with P100 filter assembly
  • Extended field of vision
  • Large inhalation valves allow for larger volume of air per breath
  • Exhalation valve is strategically positioned under the nostrils and in front of the mouth to ensure hot, exhaled air exits the mask immediately
  • Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator facepiece
  • Offset connector improves visibility and allows face shields or welding helmets to be worn without interference

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Black
Filter Type P100
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Small/Medium
With Exhalation Valve Yes
Facepiece Material Thermoplastic Elastomer
For Use With (Application) Welding, dust, oil mists
For Use With (Equipment) Cartridges/Diskits
Model 4-SM-LP0ZAM
Packaging Type Display Bag
Length (Metric) 10.16 cm
Width (Metric) 17.78 cm
Height (Metric) 12.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.