Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dentec Safety™ Series 300 Thermoplastic Facepiece Respirators with P100 Cartridges
SDP

Catalog No. 19222450 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
Size:
Medium/Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-222-450 Medium/Large
19-222-449 Small/Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-222-450 Supplier Dentec Safety™ Supplier No. 3MLLP0ZAM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High efficiency P100 thermoplastic respirators are suitable for toxic dusts, mists and fumes, lead dusts and fumes, brake dusts, exotic welding, glass fibers, and asbestos (Note: Specific limitations apply for asbestos removal, see OSHA 1910.1001).

  • Low profile Series 300 thermoplastic half-mask respirators with P100 high efficiency filters
  • Large inhalation valves allows users to inhale a larger volume of air per breath, reducing breathing resistance
  • Exhalation valve is strategically positioned under the nostrils and in front of the mouth to ensure hot, exhaled air exits the mask immediately
  • Exhalation valve diaphragm has channels that act like a parachute to traps exhaled air and lift the diaphragm off the valve seat with very little effort, creating more space for air to exit the mask, thereby reducing heat build up inside the mask
  • Rolled sealing flange provides superior comfort and improves fit
  • Textured sealing area reduces risk of the mask slipping on the face
  • Enhanced harness assembly features:
    • Metal tabs at end to prevent nylon strap fraying and allows for adjustment when wearing gloves
    • Wide nylon strap maintains elasticity longer
    • 4-point swiveling adjustments allows the mask to be adjusted to the individual features of the wearer
    • Cradle suspension fits comfortably under suspension systems for hard hats, grinding and welding helmets
  • 3-position adjustment allows users to adjust cartridge to three different positions

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Gray
Filter Type P100
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Medium/Large
With Exhalation Valve Yes
Facepiece Material Thermoplastic Elastomer
For Use With (Equipment) Cartridges/Diskits
Model 3-ML-LP0ZAM
Packaging Type Display Bag
Length (Metric) 10.16 cm
Width (Metric) 17.78 cm
Height (Metric) 12.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.