Learn More
Glass Expansion High Efficiency Quartz Torch for Elan/NexION
SDP
Catalog No. 501907145
Encompass_Preferred
High Efficiency Quartz Torch for Elan/NexION (W1008384). Compatible with PerkinElmer Elan 6000, 9000, DRC II, DRC-E and NexION 300/350 ICP-MS. Standard Quartz Torch is P/N 30-808-8107. Use GazFit Connectors for 6mm OD torch side arms PKT 4, (GAZ-06), which make connections to torch side arms with soft walled tubing for argon gas supply.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7145 Supplier Glass Expansion Supplier No. 318082784
High Efficiency Quartz Torch for Elan/NexION (W1008384). Compatible with PerkinElmer Elan 6000, 9000, DRC II, DRC-E and NexION 300/350 ICP-MS. Standard Quartz Torch is P/N 30-808-8107. Use GazFit Connectors for 6mm OD torch side arms PKT 4, (GAZ-06), which make connections to torch side arms with soft walled tubing for argon gas supply.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001