Learn More
Bachem H-Homopro-OH
SDP
Supplier:  Bachem 4006540.0025
Encompass_Preferred
H-Homopro-OH, 3105-95-1, C6H11NO2, Mr 129.16, 25g. L-Pipecolic acid, H-Pip-OH. High enantioselectivies could be achieved (> 98% ee) when choosing L-pipecolic acid as catalyst for asymmetric Mannich reactions.
Catalog No. 50-194-7204