Learn More
New England Biolabs, Inc. HiFi Taq DNA Ligase – 50 reactions
SDP
Catalog No. NC1188720
Encompass
An optimized blend of a thermostable DNA Ligase and a proprietary additive, HiFi Taq DNA Ligase efficiently seals nicks in DNA with unmatched high fidelity. The formation of a phosphodiester bond between juxtaposed 5 phosphate and 3 hydroxyl termini of two adjacent oligonucleotides that are hybridized to a complementary target DNA is enhanced in the improved reaction buffer and mismatch ligation is dramatically reduced. The improved formulation allows higher resolution discrimination between ligation donors and acceptors, enabling precise detection of SNPs and other allele variants. HiFi Taq DNA Ligase is active at elevated temperatures (37-75C). Please note that HiFi Taq DNA Ligase is intended for use in molecular diagnostics applications that depend on high fidelity nick ligation. It is not a substitute for T4 DNA ligase and is not suitable for cloning applications or adapter ligation/NGS library prep.
Catalog No. NC1188720 Supplier New England Biolabs, Inc. Supplier No. M0647S
An optimized blend of a thermostable DNA Ligase and a proprietary additive, HiFi Taq DNA Ligase efficiently seals nicks in DNA with unmatched high fidelity. The formation of a phosphodiester bond between juxtaposed 5 phosphate and 3 hydroxyl termini of two adjacent oligonucleotides that are hybridized to a complementary target DNA is enhanced in the improved reaction buffer and mismatch ligation is dramatically reduced. The improved formulation allows higher resolution discrimination between ligation donors and acceptors, enabling precise detection of SNPs and other allele variants. HiFi Taq DNA Ligase is active at elevated temperatures (37-75C). Please note that HiFi Taq DNA Ligase is intended for use in molecular diagnostics applications that depend on high fidelity nick ligation. It is not a substitute for T4 DNA ligase and is not suitable for cloning applications or adapter ligation/NGS library prep.