Learn More
LAMDA BIOTECH INC High Fidelity PCR 2x Master Mix
Catalog No. 502045765
Encompass_Preferred
The Ready-to-use HFL PCR Master Mix is for high fidelity and long fragment PCR, which contains modified high fidelity thermal stable DNA polymerases in a pre-optimized PCR buffer. Applying the extra high fidelity thermal stable DNA polymerases in the master mix ensures not only the highest fidelity for the PCR product, but also improves the amplification length of the resulting DNA fragments and the robust amplification efficiency. Amplified DNA contains blunt-ended DNA fragments. The Master Mix is useful when cloning large DNA fragments at high fidelity; such as, promoter and other important DNA regions. The HFL PCR Master Mix is in the 2X format. For most of the PCR experiments with this master mix, only template, primers and H2O will be needed.
Catalog No. 50-204-5765 Supplier LAMDA BIOTECH INC Supplier No. D135HFL
The Ready-to-use HFL PCR Master Mix is for high fidelity and long fragment PCR, which contains modified high fidelity thermal stable DNA polymerases in a pre-optimized PCR buffer. Applying the extra high fidelity thermal stable DNA polymerases in the master mix ensures not only the highest fidelity for the PCR product, but also improves the amplification length of the resulting DNA fragments and the robust amplification efficiency. Amplified DNA contains blunt-ended DNA fragments. The Master Mix is useful when cloning large DNA fragments at high fidelity; such as, promoter and other important DNA regions. The HFL PCR Master Mix is in the 2X format. For most of the PCR experiments with this master mix, only template, primers and H2O will be needed.