Learn More
Apexbio Technology LLC hyPerFUsion™ high-fidelity PCR Kit, 500U
SDP
Supplier:  Apexbio Technology LLC K1033500
Encompass_Preferred
hyPerFUsion™ high-fidelity DNA polymerases are composed of a DNA-binding domain fused to a Pyrococcus-like proofreading polymerase. Because of this, hyPerFUsion™ DNA polymerases produce PCR products with very high speed and accuracy. Also, hyPerFUsion™ DNA polymerases are tolerant of various inhibitors, allowing for robust amplification with minimal optimization. hyPerFUsion™ DNA Polymerase is an ideal choice for cloning and can be used for long or difficult (GC-rich) amplicons. hyPerFUsion™ high-fidelity DNA polymerases have become the choice for even demanding applications like massively parallel high-throughput sequencing of whole genomes.High-Fidelity DNA Polymerases are important for applications in which the DNA sequence needs to be correct after amplification. Its unique structure, a novel Pyrococcus-like enzyme fused with a processivity-enhancing domain, increases fidelity and speed. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5576