Learn More
Meridian Life Science High-Fidelity Pfu, 200UL
SDP
Catalog No. 5000856244
Encompass_Preferred
High-Fidelity Pfu is a thermostable high fidelity DNA polymerase enzyme, isolated from Pyrococcus furiosus. The Pfu polymerase enzyme replicates DNA in the 5´-3´ direction in the presence of magnesium, but also possesses 3´-5´ exonuclease (proofreading) activity, making it ideal for NGS library amplification.
Catalog No. 50-008-56244 Supplier Meridian Life Science Supplier No. MDX00302
High-Fidelity Pfu is a thermostable high fidelity DNA polymerase enzyme, isolated from Pyrococcus furiosus. The Pfu polymerase enzyme replicates DNA in the 5´-3´ direction in the presence of magnesium, but also possesses 3´-5´ exonuclease (proofreading) activity, making it ideal for NGS library amplification.