Learn More
Chemglass Life Sciences High Flow Manifold System with Hard Connect Fittings, 75L-150L Reactor Capacity, M30 Circulator Size End Fitting, 1-1/2" Beaded Pipe Triple Insulated Flexible Hose, -60 to 200°C Temperature Range
SDP
Catalog No. 502733546
Encompass_Preferred
The Chemglass High Flow Manifold System provides a safe, low-stress connection between a circulator and a glass process reactor jacket. Designed for 75-150 L reactor jackets, the manifold reduces mechanical strain on glass inlet/outlet ports and supplies pressure monitoring, relief, and hose connections for jacket fluid circulation. Hoses to connect the manifold to an external circulator are not supplied and must be ordered separately.
  • Suitable for 75-150 L reactor jackets.
  • Hard connect fittings reduce mechanical stress on glass inlet and outlet ports.
  • Includes triple insulated flexible hoses with welded stainless steel beaded-pipe fitting on one end and 3/4 in male NPT on the other.
  • Provides a pressure gauge (0-15 psi) and a pressure relief valve preset and safety sealed to 12 psi.
  • Includes a stainless steel ball valve for draining jacket fluid.
  • Operates over -60 to 200 C.
Catalog No. 50-273-3546 Supplier Chemglass Life Sciences Supplier No. CG1969M09
The Chemglass High Flow Manifold System provides a safe, low-stress connection between a circulator and a glass process reactor jacket. Designed for 75-150 L reactor jackets, the manifold reduces mechanical strain on glass inlet/outlet ports and supplies pressure monitoring, relief, and hose connections for jacket fluid circulation. Hoses to connect the manifold to an external circulator are not supplied and must be ordered separately.
  • Suitable for 75-150 L reactor jackets.
  • Hard connect fittings reduce mechanical stress on glass inlet and outlet ports.
  • Includes triple insulated flexible hoses with welded stainless steel beaded-pipe fitting on one end and 3/4 in male NPT on the other.
  • Provides a pressure gauge (0-15 psi) and a pressure relief valve preset and safety sealed to 12 psi.
  • Includes a stainless steel ball valve for draining jacket fluid.
  • Operates over -60 to 200 C.