Learn More
Chemglass Life Sciences High Flow Manifold System with Hard Connect Fittings, 10L-50L Reactor Capacity, M30 Circulator Size End Fitting, 1" Beaded Pipe Triple Insulated Flexible Hose, -60 to 200°C Temperature Range
SDP
Catalog No. 50872177
Encompass_Preferred
High Flow Manifold System with Hard Connect Fittings, 10L-50L Reactor Capacity, M30 Circulator Size End Fitting, 1" Beaded Pipe Triple Insulated Flexible Hose, -60 to 200C Temperature Range. System is supplied complete as illustrated with Triple Insulated Flexible Hose. Hoses have welded on S.S. Beaded Pipe fitting on one end and S.S. 3/4 Male NPT fitting on the other. Hoses to connect manifold to circulator are NOT supplied and must be ordered separately.Inlet Features: Triple Insulated Flexible Hose S.S. Ball Valve For Draining Jacket FluidOutlet Features: 2" Pressure Gauge, 0-15 PSI Pressure Relief Valve Preset And Safety Sealed To 12 PSI 3/4 ID Triple Insulated Flexible Hose Has Welded On S.S. Beaded Pipe Fitting For Direct Connection To Inlet/OutletCG-1969-M-04 Fits Support Frame CG-1965,CG-1965-X, and CG-1968-FPLEASE NOTE: MANIFOLDS WITH QUICK CONNECT FITTINGS ARE NOT RECOMMENDED FOR USE WITH HUBER CIRCULATORS WITH M30 FITTINGS.
Catalog No. 50-872-177 Supplier Chemglass Life Sciences Supplier No. CG1969M04
High Flow Manifold System with Hard Connect Fittings, 10L-50L Reactor Capacity, M30 Circulator Size End Fitting, 1" Beaded Pipe Triple Insulated Flexible Hose, -60 to 200C Temperature Range. System is supplied complete as illustrated with Triple Insulated Flexible Hose. Hoses have welded on S.S. Beaded Pipe fitting on one end and S.S. 3/4 Male NPT fitting on the other. Hoses to connect manifold to circulator are NOT supplied and must be ordered separately.Inlet Features: Triple Insulated Flexible Hose S.S. Ball Valve For Draining Jacket FluidOutlet Features: 2" Pressure Gauge, 0-15 PSI Pressure Relief Valve Preset And Safety Sealed To 12 PSI 3/4 ID Triple Insulated Flexible Hose Has Welded On S.S. Beaded Pipe Fitting For Direct Connection To Inlet/OutletCG-1969-M-04 Fits Support Frame CG-1965,CG-1965-X, and CG-1968-FPLEASE NOTE: MANIFOLDS WITH QUICK CONNECT FITTINGS ARE NOT RECOMMENDED FOR USE WITH HUBER CIRCULATORS WITH M30 FITTINGS.