Learn More
Waters Corp Waters Corporation High Flow Nebulizer, 2424 ELS, 300-3000 μL/min
Catalog No. 50791683 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This high flow nebulizer is a standard component designed for the 2424 Evaporative Light Scattering (ELS) detector. Its primary function is to mix chromatographic effluent with a carrier gas, typically nitrogen, to create an aerosol of fine droplets for entry into a narrow-orifice drift tube.
  • Designed for flow rates ranging from 300 to 3000 μL/min.
  • Features a variable control heater to warm sample solution and improve mass flow into the drift tube.
  • Cooling capability for faster equilibration times and enhanced sensitivity of semi-volatile samples.
Catalog No. 50-791-683 Supplier Waters Corp Supplier No. 205000458
This high flow nebulizer is a standard component designed for the 2424 Evaporative Light Scattering (ELS) detector. Its primary function is to mix chromatographic effluent with a carrier gas, typically nitrogen, to create an aerosol of fine droplets for entry into a narrow-orifice drift tube.
  • Designed for flow rates ranging from 300 to 3000 μL/min.
  • Features a variable control heater to warm sample solution and improve mass flow into the drift tube.
  • Cooling capability for faster equilibration times and enhanced sensitivity of semi-volatile samples.