Learn More
Perkin Elmer US LLC High Flow ST nebulizer line, connects to port #3 0.25 i.d. (green marker)
Catalog No. 509047740
Encompass_Preferred
The FAST is a high-throughput automated sample introduction system for PerkinElmer ICP-OES and ICP-MS. The FAST increases productive instrument time by reducing sample uptake, stabilization and rinse steps, allowing you to run more samples each day. Features and Benefits Sample never touches peristaltic pump tubing FAST uptake, stabilization and washout P-Series valve and loop combination has superb washout Carrier stream without segmented air bubbles ensures high-precision sample introduction Direct integration of MP2 peripump and FAST valve with instrument.  
Catalog No. 50-904-7740 Supplier Perkin Elmer US LLC Supplier No. N0777287
The FAST is a high-throughput automated sample introduction system for PerkinElmer ICP-OES and ICP-MS. The FAST increases productive instrument time by reducing sample uptake, stabilization and rinse steps, allowing you to run more samples each day. Features and Benefits Sample never touches peristaltic pump tubing FAST uptake, stabilization and washout P-Series valve and loop combination has superb washout Carrier stream without segmented air bubbles ensures high-precision sample introduction Direct integration of MP2 peripump and FAST valve with instrument.  