Learn More
Cole-Parmer Cole-Parmer High-Form Crucible with Cover, Porcelain; 50 mL
Catalog No. NC0733140 Shop All Cole-Parmer Products
Encompass_Preferred
High-form crucibles are glazed inside and out except for outside bottom. Withstands temperatures to 2102°F (1150°C). Cover is included.
Catalog No. NC0733140 Supplier Cole-Parmer Supplier No. 6310052
High-form crucibles are glazed inside and out except for outside bottom. Withstands temperatures to 2102°F (1150°C). Cover is included.